missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Filtration Hardware, Accessories, and Spare Parts
GREENER_CHOICE

Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.

Brand:  Thermo Scientific™ DS0390-0070

26.29 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10634212

  • £41.25 / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.

Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Polycarbonate
44.8 in. (with Adaptor)
2.75 in.
Overal height w/adapter: 114mm; Bell I.D.: 70mm
Filling Bell
114 mm (with Adaptor)
70 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.