missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Polycarbonate Safety Splash Shield
79.77 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Use promo code "23291" to get your promotional price.
Click to view available options
Description:
Safety Splash Shield with acetal flexible arm; PC 21.6 x 27.9 cm
Length (English):
12 in.
Width (English):
8.5 in.
Description
Simultaneously maintain safety and a clear field of vision by using the Thermo Scientific™ Nalgene™ Polycarbonate Safety Splash Shield. This shield attaches easily to benchtop shelving, machinery or other magnetic surfaces for convenient protection from splashes, aerosols and flying debris. This durable, scratch-resistant 1/8″ (3mm) thick polycarbonate shield protects the user and features an easy to maneuver arm. Meets OSHA 29CFR1910.1030 standard.
- Flexible 14 inch arm makes it easy to maneuver shield over samples
- Magnetic base firmly grips to magnetic surfaces letting you position the shield horizontally (flat), vertically or on an overhead surface
- Provides UV protection from 200-360nm for added versatility
- Biohazard/Transparent
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Certifications/Compliance | Meets OSHA 29CFR1910.1030 standard |
Thickness (English) | 0.12 in. |
Thickness (Metric) | 3 mm |
Width (English) | 8.5 in. |
Width (Metric) | 21.5 cm |
Description | Safety Splash Shield with acetal flexible arm; PC 21.6 x 27.9 cm |
Lens Tint | Clear |
Product Type | Safety Splash Shield |
Length (English) | 12 in. |
No. per Case | 4/Cs. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'