missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Polycarbonate, Validation Bottle
Click to view available options
Capacity (Metric):
30 mL
250 mL
2 L
Description
Perform QA lot sample storage of high-valued pharmaceuticals using Thermo Scientific™ Nalgene™ Polycarbonate Validation Bottles. Small-volume bottles are molded of the same materials as larger carboys, but require a lower volume of expensive reagents to perform validation testing.
Autoclavable, transparent polycarbonate construction allows for convenient material compatibility validation.
Includes:
White polypropylene screw closure and TPE gasket

Specifications
Specifications
Autoclavable | Autoclavable |
Capacity (English) | 0.5 gal. |
Capacity (Metric) | 2 L |
Closure Material | Polypropylene |
Closure Size | 53B |
Color | Clear |
Diameter (Metric) Inner Neck | 42 mm |
Diameter (Metric) Outer | 123 mm |
For Use With (Application) | Perform validation testing |
Height (Metric) With Closure | 296 mm |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1404393 | Certificate of Compliance | 07/04/2024 | DS2127-2000 |
1377545 | Certificate of Compliance | 09/01/2023 | DS2127-2000 |
1304959 | Certificate of Compliance | 05/02/2021 | DS2127-2000 |
1270714 | Certificate of Compliance | 20/11/2019 | DS2127-2000 |
1247143 | Certificate of Compliance | 16/01/2019 | DS2127-2000 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Nalgene™ Polycarbonate, Validation Bottle >
Spot an opportunity for improvement?Share a Content Correction