missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polyetherimide Replacement Vacuum Jars
GREENER_CHOICE

Perform demanding applications with these chemically resistant replacement vacuum jars, which weigh less than glass and have a warm amber cast.

Brand:  Thermo Scientific™ DS5320-1212

 View more versions of this product

Product Code. 10719041

  • £424.00 / Each
Estimated Shipment: 17-03-2025
to see stock.
Explore more special offers
Add to basket

Alert:

Do not autoclave vacuum jars when used for vacuum service. Do not use with unsaturated halogenated hydrocarbons.

This item is not returnable. View return policy

Description

Description

Perform demanding applications with chemically resistant Thermo Scientific™ Nalgene™ Replacement Vacuum Jars. These jars weigh less than glass and have a warm amber cast.

Features:
  • Identical to jars included with Part Nos. 5305-0910 and 5305-1212
  • Easier to handle than glass and won't break, chip or shatter
  • Transparent

Ordering Information:

Jar only; plate and gaskets must be ordered separately

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Brim Cap
5 gal.
Transparent Amber
305 mm
305 mm
Replacement Vacuum Jar
Nalgene™ Vacuum Chambers
18.9 L
12 in.
12 in.
Polyetherimide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.