Learn More
Thermo Scientific™ Nalgene™ Polyolefin Pressure-Sensitive Labels
Combine these waterproof, chemical resistant labels with photocopiers, laser printers or inkjet printers. Will not wrinkle, curl or melt.
£64.05 - £383.00
Product Code | Brand | Height (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
11333482
|
Thermo Scientific™
6319-0010 |
22 mm | 22 mm |
£262.45
Pack of 4000 |
In Stock
Log in to see stock. |
||||
10713052
|
Thermo Scientific™
6319-0015 |
20 mm | 38 mm |
£371.00
Pack of 3250 |
In Stock
Log in to see stock. |
||||
11319821
|
Thermo Scientific™
6319-0020 |
25 mm | 67 mm |
£64.05
Pack of 300 |
In Stock
Log in to see stock. |
||||
11394555
|
Thermo Scientific™
6319-0030 |
13 mm | 44 mm |
£383.00
Pack of 4000 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
11353482
|
Thermo Scientific™
6319-0040 |
51 mm | 102 mm |
£262.45
Pack of 500 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
Description
Combine Thermo Scientific™ Nalgene™ Polyolefin Pressure-Sensitive Labels with photocopiers, laser printers or inkjet printers. Waterproof and compatible with most lab chemicals, Nalgene polyolefin labels will not wrinkle, curl or melt. They accept a variety of inks including copier, marker, ballpoint, gels and technical markers.
Durable Polyolefin Labels
- Rated to −29°C
- Resistant to ultraviolet light, mildew, chemicals and dirt and won't discolor, crack or tear
- Excellent for labeling microscopic slides— adhesive backing sticks well to any plastic, glass or metal surface
- Chemically compatible with most lab reagents although solvents may dissolve adhesive
- Autoclavable
- Formatted for easy setup in word processing programs; instructions included in each pack
Note: For use with all writing devices except pencil. Test specific inks for suitability prior to use.
.
Specifications
Label | |
Polyolefin |