Learn More
Thermo Scientific™ Nalgene™ Polyolefin Right-To-Know Custom Labeling System
Create labels that identify a chemical and its properties using the Thermo Scientific™ Nalgene™ Polyolefin Right-To-Know Custom Labeling System.
Brand: Thermo Scientific™ 6316-1000
57.13 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.12960kg
Description
Create labels that identify a chemical and its properties using the Thermo Scientific™ Nalgene™ Polyolefin Right-To-Know Custom Labeling System.
Features:- Overlay provided to protect label from chemical contact
- Complete instructions included with each package
- Hazard symbol stickers are GHS-compliant
Specifications
Custom Right-To-Know Label System | |
Label | |
150 | |
5 in. | |
3 in. |
Assorted | |
Polyolefin | |
25 | |
127 mm | |
76.2 mm |