missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels
GREENER_CHOICE

Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.

Brand:  Thermo Scientific™ 4250-0075

31.72 GBP valid until 2024-11-29
Use promo code "21718" to get your promotional price.


 View more versions of this product

Product Code. 10294491

  • £38.25 / Pack of 6
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.

  • Body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

76 mm
7 mm
Autoclavable
80 mm
114 mL
Polypropylene
Funnel Analytical PP 75mm
6/Pk.; 36/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers