Learn More
Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels
Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.
Brand: Thermo Scientific™ 4250-0065
43.02 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : A0
Additional Details : Weight : 0.19350kg
Description
Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.
- Body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration
- Autoclavable
Specifications
63 mm | |
7 mm | |
Yes | |
58 mm |
64 mL | |
Polypropylene | |
Funnel Analytical PP 65mm | |
12/Pk.; 36/Cs. |