missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels
GREENER_CHOICE

Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.

Brand:  Thermo Scientific™ 4250-0160

32.14 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10505981

  • £44.55 / Pack of 2
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.

  • Body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

158 mm
13 mm
Autoclavable
152 mm
976 mL
Polypropylene
Funnel Analytical PP 160mm
2/Pk.; 12/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.