missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Autoclaving Baskets
GREENER_CHOICE

Hold labware during autoclaving and washing with these polypropylene autoclaving baskets, which also allow for complete venting and draining.

Brand:  Thermo Scientific™ 6917-0127

35.46 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.


 View more versions of this product

Product Code. 10676541

  • £51.15 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Hold labware during autoclaving and washing with Thermo Scientific™ Nalgene™ Polypropylene Autoclaving Baskets, which also allow for complete venting and draining. These baskets won't corrode and are chemically compatible with most acids, bases and alcohols. The smooth surface will not scratch plastic or glass contents.

  • Nestable for storage
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polypropylene Autoclaving Basket
Autoclavable
Polypropylene
6
6.125 x 4.87 x 4.125 in. (15.6 x 10.4 x 10.4cm)
6.125 in.
4.87 in.
4.125 in.
Autoclaving Basket PP 105 x 123 x 154mm
White
Square
1
15.6 cm
10.4 cm
10.4 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.