missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene, Carboy

Product Code. 10413491
Click to view available options
Capacity (Metric):
10 L
20 L
50 L
Unit Size:
Each

Product Code. 10413491

Brand: Thermo Scientific™ 22500020

Only null left
Add to basket
Edge
Add to basket

For Use With: For use with large batches of culture media, vaccine product production, bulk media and chemical storage; Autoclavable; Closure Material: Polypropylene; Color: White

Choose Thermo Scientific™ Nalgene™ Autoclavable Carboys for large batches of culture media, vaccine product production, bulk media and chemical storage. These carboys feature a large mouth for easy filling/dispensing, and are made of natural polypropylene with white 83B polypropylene closure and TPE (thermoplastic elastomer) gasket.

Customization options available for fluid transfer applications. Fit perfectly with a variety of optional 83B closures with standard tubing sets (such as Nalgene™ Top Works™, Nalgene™ Filling/Venting) and custom tubing set designs.

Key Features:

  • Convenient shoulder handles allow easy carrying and pouring, even with gloved hands
  • Available in 10, 20, and 50 L sizes for a variety of biopharmaceutical applications
  • Graduated to contain in 1 gal. and 5 L increments for ease of use
  • Nalgene Top Works Fluid Transfer Systems (Cat. No. 2135) permits convenient liquid aseptic transfer
  • Autoclavable/Graduated/Leakproof†

Ordering Information:

  • Additional closure options are Nalgene Quick Filling/Venting Closures (Cat. No. 2158) or Nalgene Filling/Venting Closures (Cat. No. 2162)
  • Additional accessories include Nalgene Barbed Bulk Head Fittings (Cat. No. 6149)
  • Custom Tubing Sets are available-please refer to Thermo Scientific Custom Tubing Set Configurator at www.thermoscientific.com/tubingconfigurator.com
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Capacity (English) 2.6 gal.
Capacity (Metric) 10 L
Closure Material Polypropylene
Closure Size 83B
Color White
Diameter (Metric) Outer 250 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
Height (Metric) With Closure 389 mm
Includes Handles
Material Polypropylene
No. per Pack 1 Each
Product Type Carboy with Handle
Sterility Non-sterile
Volume (English) Brim 3 gal.
Volume (Metric) Brim 12 L
Workflow Step Upstream media/buffer prep and storage. Upstream process solution storage
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.