missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
GREENER_CHOICE

Combine these flasks with most lab reagents and solutions due to their excellent chemical compatibility and shatter-proof construction.

Brand:  Thermo Scientific™ 4102-2000

39.04 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10065200

  • £121.00 / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Choose Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks for use with a wide range of lab reagents and solutions due to their excellent chemical compatibility. Rugged, shatterproof construction and light weight compared with glass make these flasks a valuble element of lab safety programs.

  • Translucent wall makes liquid level easily visible
  • 2000mL size has molded-in graduations
  • Mouth sized to accept standard sized stoppers (stopper sold separately)
  • Repeatedly autoclavable for long life and rigorous use
TRUSTED_SUSTAINABILITY
Specifications

Specifications

64 oz.
2000 mL
6
Autoclavable
Yes
Non-sterile
Stopper No. 10
Flask Erlenmeyer PP 2000mL
PPCO
2
Reusable
Narrow
Plain
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers