Learn More
Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
Combinethese flasks with most lab reagents and solutions due to their excellent chemical compatibility and shatter-proof construction.
Brand: Thermo Scientific™ 4102-1000
25.38 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 55
Additional Details : Weight : 1.06200kg
Description
Choose Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks for use with a wide range of lab reagents and solutions due to their excellent chemical compatibility. Rugged, shatterproof construction and light weight compared with glass make these flasks a valuble element of lab safety programs.
- Translucent wall makes liquid level easily visible
- 2000mL size has molded-in graduations
- Mouth sized to accept standard sized stoppers (stopper sold separately)
- Repeatedly autoclavable for long life and rigorous use
Specifications
32 oz. | |
1000 mL | |
6 | |
Autoclavable | |
No | |
Non-sterile | |
Stopper No. 8 |
Flask Erlenmeyer PP 1000mL | |
PPCO | |
2 | |
Reusable | |
Narrow | |
Plain |