missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels
GREENER_CHOICE

Transfer powders with these powder funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. A shatter-proof alternative to glass.

Brand:  Thermo Scientific™ 4252-0150

32.72 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10306281

  • £61.50 / Pack of 4
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Transfer powders with Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. This shatter-proof alternative to glass is chemical resistant to a wide range of lab chemicals and reagents and can make a valuble contribution to lab safety programs.

  • Polypropylene is break-resistant and chemically compatible with a wide variety of lab reagents
  • Suitable for general lab use, it is a durable replacement for glass funnels
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

145 mm
29 mm
137 mm
Funnel Powder PP 150mm
4/Pk.; 24/Cs.
716 mL
Polypropylene
Autoclavable
32 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.