missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels
GREENER_CHOICE

Product Code. p-4525012
Click to view available options
Diameter (Metric)Top:
64 mm
79mm
100 mm
145 mm
Capacity (Metric):
68 mL
115 mL
243 mL
716 mL
Unit Size:
Pack of 12
Pack of 4
Pack of 6

Product Code. 10545211

Brand: Thermo Scientific™ 42520065

Only null left
Add to basket
Edge
Add to basket

Transfer powders with these powder funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. A shatter-proof alternative to glass.

Transfer powders with Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. This shatter-proof alternative to glass is chemical resistant to a wide range of lab chemicals and reagents and can make a valuble contribution to lab safety programs.

  • Polypropylene is break-resistant and chemically compatible with a wide variety of lab reagents
  • Suitable for general lab use, it is a durable replacement for glass funnels
  • Autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric)Top 64 mm
Capacity (Metric) 68 mL
Diameter (Metric) Stem 17 mm
Material Polypropylene
Height (Metric) 67 mm
Autoclavable Autoclavable
Description Funnel Powder PP 65mm
Length (Metric) Stem 22mm
Quantity 12/Pk.; 36/Cs.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.