Learn More
Thermo Scientific™ Nalgene™ Polypropylene Scissor-Type Forceps
Avoid corrosion from acid exposure with Thermo Scientific™ Nalgene™ Polypropylene Scissor-Type Forceps, which are chemically compatible with most lab reagents.
Brand: Thermo Scientific™ 6320-0010
34.68 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 55
Additional Details : Weight : 0.06750kg
Description
Avoid corrosion from acid exposure with Thermo Scientific™ Nalgene™ Polypropylene Scissor-Type Forceps, which are chemically compatible with most lab reagents. Double-action hinges provide extra leverage and long life. These forceps feature a scissor-type handle with a ratchet to lock tips shut and a handy clamp for chromatography columns.
- Autoclavable
- Feature jaws with interlocking teeth to grip objects tightly
Specifications
Straight, Serrated | |
Polypropylene | |
Forceps | |
Scissor Type |
12 | |
Forceps | |
12/Pk. |