Learn More
Replacement Stopcock Assembly for Nalgene™ Polypropylene Separatory Funnels with Closure
Perform a variety of lab duties with chemically inert, translucent and autoclavable Thermo Scientific™ Nalgene™ Polypropylene Separatory Funnels.
Brand: Thermo Scientific Nalgene 714301-1025
83.21 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.10000kg
Description
This is a replacement part for Nalgene™ 4300- and 4301- Separatory Funnels.
Perform a variety of lab duties with chemically inert, translucent and autoclavable Thermo Scientific™ Replacement Stopcock Assembly for Nalgene™ Polypropylene Separatory Funnels with Closure. Closure has excellent chemical resistance and requires no lubrication.
Features:
- Separation lines between immiscible liquids are easily observed
- Phase interface is observable down to the stopcock
- TFE stopcocks need no lubrication, allowing for easier handling and operation
- Funnel is chemically compatible with a wide range of lab reagents
- Autoclavable/Leakproof
Includes:
- Polypropylene funnel, TFE stopcock and housing, polypropylene screw closure
- For replacement stopcock, order Cat. No. 714301-1025
- Funnels are susceptible to softening and swelling from extended exposure to aggressive solvents though the effect is reversible.
- Stem lengths, 65mm. Autoclavable except for stopcock assembly, which is easily removed for chemical sterilization.
- Autoclavable except for stopcock assembly, which is easily removed and chemically sterilized.
Specifications
TFE | |
Replacement Teflon Stopcock Assembly | |
Nalgene™ 4300- and 4301- Separatory Funnels |