Learn More
Thermo Scientific™ Nalgene™ PPCO Conical-Bottom Centrifuge Tubes
Simplify measurements using these PPCO conical-bottom centrifuge tubes, which feature molded-in graduations. Translucent PPCO has excellent chemical resistance.
£53.35 - £56.02
Specifications
Material | Polypropylene Copolymer |
---|---|
No. per Case | 100 |
No. per Pack | 10 |
Sterility | Non-sterile |
Autoclavable | Autoclavable |
Product Code | Brand | Diameter (Metric) | Capacity (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Capacity (Metric) | Length (Metric) | Price | Quantity & Availability | |||
10374061
|
Thermo Scientific™
3103-0015 |
16.8 mm | 15 mL | 118.6 mm |
£53.35
Pack of 10 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||
10602682
|
Thermo Scientific™
3103-0050 |
28.7 mm | 50 mL | 133.1 mm |
£56.02
Pack of 10 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||
Description
Simplify measurements using Thermo Scientific™ Nalgene™ PPCO Conical-Bottom Centrifuge Tubes, which feature molded-in graduations. Translucent polypropylene copolymer (PPCO) has excellent chemical resistance.
Features:- Translucent Polypropylene Copolymer (PPCO) is compatible with a wide range of lab reagents
- Designed for low-speed centrifugation up to 6000 × g in refrigerated or non-refrigerated centrifuges (Tubes must be filled at least 80% for proper performance)
- Conical bottoms concentrate pellet in a small area for easy isolation and retrieval
- Molded-in graduations last the life of the tube
- Last longer than polycarbonate tubes under conditions of repeated autoclaving
- Tube are both autoclavable and graduated
- Autoclavable
Ordering Information:
Supplied without closure; friction fit closures available. For 15mL use DS3111-00017; for 50mL use DS3111-0029.
Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) centrifuge tubes are designed to last many spins under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. These centrifuge tubes are recyclable in many communities. At end of product life, please wash and recycle your Nalgene PP and PPCO centrifuge tubes if services are available in your community.
Specifications
Polypropylene Copolymer | |
10 | |
Autoclavable | |
Conical | |
Molded-in | |
6000 x g |
100 | |
Non-sterile | |
Conical | |
No | |
Centrifuge Tube |