Learn More
Thermo Scientific™ Nalgene™ PPCO Low-Profile Closures for Micro Packaging Vials: Nonsterile, Bulk Pack
Identify samples quickly with low-profile, color-coded closures molded of high-purity, low metal content PPCO resin.
Brand: Thermo Scientific™ 362821-0115
76.00 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : A0
Additional Details : Weight : 0.39300kg
Description
Quickly identify samples using Thermo Scientific™ Nalgene™ PPCO Low-Profile Micro Packaging Vial Closures, offered in a variety of colors. Closures are molded of high-purity, low metal-content polypropylene copolymer resin—excellent for PCR reagents.
Features:
- Vials (listed separately) and closures are pressure-tested together at 7.5 PSIG for air shipment
- Nonsterile
- Noncytotoxic and nonpyrogenic
- Leakproof† when used with Nalgene Micro Packaging Vials
Ordering Information:
Micro Packaging Vial Closures are bulk packed without vials, 500 per inner bag, 1,000 per case. Order nonsterile vials separately (Cat. Nos. 362800, 362805).
Specifications
Low-profile closures for micro packaging vials | |
11 mm | |
Non-sterile | |
500 | |
PPCO | |
ISO 13485:2016 | |
11 mm |
Red | |
PPCO | |
1000 | |
Nalgene | |
500 Bag, 1000 Cs. | |
Closure |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.