Learn More
Thermo Scientific™ Nalgene™ PPCO Low-Profile Closures for Micro Packaging Vials: Sterile, Bulk Pack

Identify samples quickly with sterile, low-profile closures molded of high-purity low metal content PPCO resin, available in a variety of colors.
£101.36 - £114.00
Specifications
Diameter (Metric) Outer | 11 mm |
---|---|
Material | PPCO |
Sterility | Sterile |
Closure Material | PPCO |
Quantity | 500 Bag, 1000 Cs. |
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
10512425
![]() |
Thermo Scientific™
342821-0110 |
Natural |
£114.00
Pack of 1000 |
In Stock
Log in to see stock. |
||||||
11314475
![]() |
Thermo Scientific™
342821-1112 |
Pink |
£101.36
Pack of 1000 |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||||
10600914
![]() |
Thermo Scientific™
342821-0111 |
White |
£101.36
Pack of 1000 |
Estimated Shipment: 27-06-2025
Log in to see stock. |
||||||
10747283
![]() |
Thermo Scientific™
342821-0112 |
Yellow |
£101.36
Pack of 1000 |
Estimated Shipment: 18-08-2025
Log in to see stock. |
||||||
10671674
![]() |
Thermo Scientific™
342821-0114 |
Green |
£101.36
Pack of 1000 |
Estimated Shipment: 24-09-2025
Log in to see stock. |
||||||
10542615
![]() |
Thermo Scientific™
342821-0115 |
Red |
£101.36
Pack of 1000 |
Estimated Shipment: 18-08-2025
Log in to see stock. |
||||||
10384003
![]() |
Thermo Scientific™
342821-0116 |
Blue |
£101.36
Pack of 1000 |
Estimated Shipment: 19-06-2025
Log in to see stock. |
||||||
10120323
![]() |
Thermo Scientific™
342821-0118 |
Purple |
£101.36
Pack of 1000 |
Estimated Shipment: 18-08-2025
Log in to see stock. |
||||||
10344053
![]() |
Thermo Scientific™
342821-1111 |
Amber |
£114.00
Pack of 1000 |
Estimated Shipment: 27-06-2025
Log in to see stock. |
||||||
Description
Identify samples quickly using Thermo ScientificTM NalgeneTM PPCO Low-Profile Closures for Micro Packaging Vials, offered in a variety of colors. Closures are molded of high-purity, low metal-content polypropylene copolymer (PPCO) resin—excellent for PCR reagents. Closures are sterile and ready to use.
PPCO Closures:- Sterile to 10-6 SAL
- RNase/DNase free
- Non-cytotoxic and non-pyrogenic
- Leakproof† when used with Nalgene Micro Packaging Vials
Ordering Information:
Micro Packaging Vial Closures are bulk packed without vials, 500 per inner bag, 1,000 per case. Order sterile vials separately (Cat. Nos. 342800, 342805).
Specifications
11 mm | |
Sterile | |
500 Bag, 1000 Cs. | |
11 mm |
PPCO | |
PPCO | |
Closure |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.