missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes

Achieve fast flow rates and high throughput for increased filtration efficiency with Nalgene Rapid-Flow sterile disposable bottle top filters.

Brand:  Thermo Scientific™ 595-4520

79.35 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.



Code : 54

Additional Details : Weight : 1.41300kg

 View more versions of this product

Product Code. 10300461

  • £152.00 / Pack of 12
In Stock
to see stock.
Add to basket

Alert:

CAUTION: Use only sterile bottles designed for use with vacuum. Always use safety shield during vacuum procedures.


Description

Description

Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ single use bottle tops feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid-Flow bottle tops are the best choice for the filtration of cell culture media, serum, additives and buffers.

  • Available in PES and SFCA membranes. Devices are color coded according to membrane.
  • Devices feature the fastest flow rate and lowest clogging of comparable alternatives
  • 0.1um PES available for mycoplasma protection
  • 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
  • Non-pyrogenic and endotoxin free - Rapid-Flow filters meet the most rigorous testing standards for any vacuum filtration unit
  • Available with Fit for Purpose Sterile documentation
Specifications

Specifications

Single-use
Polystyrene
0.2 μm
45mm
12
500 mL
75 mm
Membrane Filter
PES
Sterile
12
Bottle Top Filter
75 mm
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers