missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Rectangular Polycarbonate Clearboy™ Carboy with Spigot
104.27 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Use promo code "23291" to get your promotional price.
Click to view available options
Capacity (Metric):
9 L
20 L
Description
Easily fill and clean the Thermo Scientific™ Nalgene™ Rectangular Polycarbonate Clearboy™ with Spigot, which features a wide-mouth opening, a sturdy stainless-steel handle and space-saving rectangular design. Clear construction allows easy viewing of carboy contents. Super tough; impact resistant. Ideal for refrigerated or frozen storage of aqueous solutions, or for dispensing sterile water, aqueous reagents and media. Nalgene polycarbonate carboys can be autoclaved before refilling to eliminate bacterial growth when storing sterile solutions. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.
Features
- Molded-in graduations in 1 gallon and 5L increments for ease in dispensing
- Stainless-steel handle for ease in transporting containers
- Molded-in spigot boss provides strength and leak resistance
- A clear wall makes this carboy ideal when visual inspection of contents is necessary
- Follow autoclave instructions provided with the product for proper function; tubing adapter is not autoclavable
- Autoclavable/Transparent/Graduated
Includes:
Polypropylene spigot and screw closure
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Autoclavable | Autoclavable |
Capacity (English) | 2.4 gal. |
Capacity (Metric) | 9 L |
Closure Material | Polypropylene |
Closure Size | 100 mm |
Color | Transparent |
Description | Autoclavable Clear Rectangular Carboy w/Spigot |
Diameter (Metric) Inner Neck | 86 mm |
For Use With (Application) | Ideal for use when visual inspection of contents is necessary |
Height (Metric) With Closure | 361 mm |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'