missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Round Polycarbonate Clearboy™ Carboy with Spigot
GREENER_CHOICE

For Use With: Suitable for mixing, storing and freezing aqueous-based reagents and biologicals, especially useful for visual inspection of contents for quality.; Autoclavable; Closure Material: Polypropylene; Color: Clear

£243.00 - £310.00

Specifications

Autoclavable Autoclavable
Closure Material Polypropylene
Closure Size 83B
Color Clear
Diameter (Metric) Inner Neck 66 mm
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10722332
GREENER_CHOICE
View accessory products View Documents Promotion Details
Thermo Scientific™
2317-0020
10 L
£243.00
Each
In Stock
to see stock.
Edge
Add to basket
 
10684071
GREENER_CHOICE
View accessory products View Documents Promotion Details
Thermo Scientific™
2317-0050
20 L
£310.00
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Prepare large volume media and cultures with Thermo Scientific™ Nalgene™ Round Polycarbonate Clearboy™ with Spigot. These durable Clearboy are lighter and safer than glass, and especially useful for visual inspection of contents for quality. Ideal for refrigerated or frozen storage of aqueous solutions, or for dispensing sterile water, aqueous reagents and media. Nalgene polycarbonate carboys can be autoclaved before refilling to eliminate bacterial growth when storing sterile solutions. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.

Features:
  • Molded in graduations in 1 gal and 5L increments for ease in dispensing
  • Molded in spigot boss provides strength and leak resistance
  • Polycarbonate is glass-clear for easy visual inspection and volume measurements
  • Suitable for mixing, storing and freezing aqueous-based reagents and biologicals
  • Designed with 83mm neck to accept TopWorks and other Nalgene accessories for versatility
  • Graduated
  • Leakproof† when used with Nalgene closures
  • Autoclavable for sterile applications 

Includes:

Polypropylene spigot screw closure and TPE gasket. See autoclaving instructions for more information.

Note:

Completely disengage closure threads and disassemble spigot before autoclaving
Follow product insert instructions for autoclaving to ensure proper function
Note: Tubing adaptor is not autoclavable
Repeated autoclaving will shorten the life of polycarbonate carboys
Use pH neutral detergent and rinse completely prior to autoclaving

Please recycle the corrugate cartons and LDPE bags in which your Nalgene carboy products are packaged.

Specifications

Specifications

Autoclavable
83B
66 mm
Molded-in graduations in 1 gal. and 5 L increments
Polycarbonate
1 Ea.
Polypropylene
Clear
Suitable for mixing, storing and freezing aqueous-based reagents and biologicals, especially useful for visual inspection of contents for quality.
Spigot
4/Cs.
Carboy with Spigot
Videos
SDS
Documents

Documents

Product Certifications

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.