Learn More
Thermo Scientific™ Nalgene™ Sealing Caps for Oak Ridge Centrifuge Tubes; PP screw closure, silicone gasket
GREENER_CHOICE
Ensure leakproof service at their maximum speeds >10,000 x g, even when full
Brand:  Thermo Scientific™ 713132-0063
25.60 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10420533
  • £40.15 / Pack of 10
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polypropylene closures with silicone gasket
Before autoclaving, set closure on top of the container without engaging the threads.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Silicone
Silicone
Replacement O-ring (for Mfr. No. DS3132-0063)