missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Septum Closures: Sterile, Bulk Pack, 38-430mm
GREENER_CHOICEBIOPHARMA

Product Code. p-4521549
Click to view available options
Neck Finish:
24-415
38-430
Unit Size:
Pack of 200
Pack of 240
This item is not returnable. View return policy

Product Code. 13127673

Brand: Thermo Scientific™ 3421780384

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Combine elastomeric sealing of silicone with nonreactive PTFE reagent contact surfaces in these septum closures for media bottles.

Combine the elastomeric sealing of silicone with nonreactive polytetrafluoroethylene reagent contact surfaces in Thermo Scientific™ Nalgene™ Septum Closures, for use with Nalgene™ Sterile PET & PETG media bottles.

Features:
  • Sterile 10-6 SAL
  • No adhesives used to attach septum to high density polyethylene closure
  • Non-cytotoxic and non-pyrogenic
  • Dual septum material for improved gas barrier

Ordering Information:

  • Closures are bulk packed for easy handling.
  • Order Nalgene PET or PETG bottles without closures separately.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description Nalgene Closure for Media Bottles
Material HDPE
No. per Case 240
No. per Pack 240
Packaging Bulk
Sterility Sterile
Liner Material Silicone, PTFE
Neck Finish 38-430
Certifications/Compliance Manufactured in compliance with ISO 13485 quality requirements
Fit for Purpose Sterile
Product Type Septum Closure
For Use With (Equipment) Nalgene Sterile PET and PETG media bottles
Closure Size 38-430 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.