Learn More
Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom: Sterile
Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing and storage.
Brand: Thermo Scientific™ 4116-1000
73.39 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Save preparation time and avoid contamination risk by using Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
- Sterile and single use to reduce contamination risk
- Collapses when autoclaved to reduce bio hazardous waste
- Graduated for quick volume assessment
- Excellent validation support: Validation binder available upon request†
Note:
† Validation binders are available to customers as support documentation during your product validation process. This confidential information can be requested at: rocregsupport@thermofisher.com. Following your request submission, a Confidentiality Agreement will be forwarded to you for completion.

Specifications
32 oz. | |
PETG | |
1 | |
None | |
Single-use | |
Narrow | |
Baffled | |
45-430 mm |
1000 mL | |
6 | |
Erlenmeyer | |
Not Autoclavable | |
Yes | |
Sterile | |
Vented |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1305750 | Certificate of Processing | 04/12/2024 | 4116-1000 |
1397225 | Certificate of Processing | 04/12/2024 | 4116-1000 |
1408571 | Certificate of Processing | 04/12/2024 | 4116-1000 |
1274453 | Certificate of Processing | 04/12/2024 | 4116-1000 |
1360983 | Certificate of Processing | 04/12/2024 | 4116-1000 |
Special Offers
-
Multiple Manufacturers
New Offer Shop Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.