missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom: Sterile

Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing and storage.

Brand:  Thermo Scientific™ 4112-2800

95.33 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.


 View more versions of this product

Product Code. 10228732

  • £143.00 / Pack of 4
In Stock
to see stock.
Explore more special offers
Add to basket

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

Save preparation time and avoid contamination risk with Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.

Features:
  • Sterile and single use: Reduce contamination risk
  • Collapses when autoclaved: Reduce bio hazardous waste
  • Graduated for quick volume assessment
TRUSTED_SUSTAINABILITY
Specifications

Specifications

95 oz.
PETG
1
None
Single-use
Narrow
Plain
70 mm
2800 mL
4
Erlenmeyer
Not Autoclavable
Yes
Sterile
Solid, non-vented
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1385597 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number4112-2800
Lot Number1331463 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number4112-2800
Lot Number1396269 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number4112-2800
Lot Number1298676 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number4112-2800
Lot Number1277807 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number4112-2800
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom: Sterile > 2800mL; Solid Closure

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.