Learn More
Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom: Sterile
Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing and storage.
Brand: Thermo Scientific™ 4115-0500
183.95 GBP valid until 2024-12-20
Use promo code "21614" to get your promotional price.
Description
Save preparation time and avoid contamination risk with Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
Features:- Sterile and single use: Reduce contamination risk
- Collapses when autoclaved: Reduce bio hazardous waste
- Graduated for quick volume assessment
Specifications
16 oz. | |
PETG | |
1 | |
None | |
Single-use | |
Narrow | |
Plain | |
45-430 mm |
500 mL | |
12 | |
Erlenmeyer | |
Not Autoclavable | |
Yes | |
Sterile | |
Vented |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.