missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure Product Code.: 10647711
Add to basket

Thermo Scientific™ Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
GREENER_CHOICE

Product Code. 10647711
1 L, 38-430 mm
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
Closure Size:
20 mm
24 mm
38-430 mm
This item is not returnable. View return policy

Product Code. 10647711

Brand: Thermo Scientific™ 20181000

Add to basket
Add to basket
This item is not returnable. View return policy

Conserve space with these square, narrow-mouth HDPE bottles that are chemically and break resistant, as well as economically priced for a wide range of lab uses.

Conserve space with Thermo Scientific™ Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure. HDPE bottles are the most chemically and break resistant of all Nalgene square bottles, yet are economically priced for a wide range of lab uses. They are made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

Features:

  • Square sides allow for good use of space and an easy grip
  • Caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Graduated for easy dispensing
  • Leakproof†
  • Can be used to -100°C (-148°F)
  • Not autoclavable
  • Participates in My Green Labs' ACT Label program

Includes:

Leakproof† Polypropylene Screw Caps

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Capacity 32 oz., 1000 mL
Capacity (Metric) 1 L
Capacity (English) 32 oz.
Color Translucent
Description Narrow-Mouth Square Bottle
Diameter (English) 3.7 in.
Diameter (English) Inner Neck 1.1 in.
Diameter (Metric) Inner Neck 28 mm
Includes Leakproof polypropylene screw closures
Material HDPE
No. per Case 24
No. per Pack 6
Sterility Non-sterile
Closure Color Natural, translucent
Graduations Yes
Certifications/Compliance USP Class VI; FDA - 21 CFR
Closure Included Yes
Autoclavable Not Autoclavable
Graduated Yes
Mouth Narrow
Shape Square
ACT Label Yes
Height (Metric) 218 mm
Product Type Square Narrow Mouth Bottle
Width (English) 3.9 in.
Height (English) 8.6 in.
Closure Material Polypropylene
Closure Size 38-430 mm
Diameter (Metric) 92 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Show More Show Less

Manuals

Package Inserts

Schematics & Diagrams

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1424901 Certificate TypeCertificate of Compliance Date02/04/2025 Catalog Number2018-1000
Lot Number1303281 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number2018-1000
Lot Number1284856 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number2018-1000
Lot Number1408983 Certificate TypeCertificate of Compliance Date25/07/2024 Catalog Number2018-1000
Lot Number1394431 Certificate TypeCertificate of Compliance Date05/03/2024 Catalog Number2018-1000

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.