Learn More
Thermo Scientific™ Nalgene™ Square Narrow-Mouth PPCO Bottles with Closure
Easily measure and dispense using these translucent, square, narrow-mouth PPCO bottles, featuring mouths wider than conventional narrow-mouth bottles.
£42.05 - £138.00
Specifications
Color | Translucent |
---|---|
Description | Narrow-Mouth Square Bottle |
Includes | Leakproof polypropylene screw closures |
Material | PPCO |
Sterility | Non-sterile |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
11784294
|
Thermo Scientific™
2016-0030 |
30 mL | 20 mm |
£42.05
Pack of 12 |
In Stock
Log in to see stock. |
||||
10044870
|
Thermo Scientific™
2016-0060 |
60 mL | 24 mm |
£50.40
Pack of 12 |
In Stock
Log in to see stock. |
||||
10054870
|
Thermo Scientific™
2016-0125 |
125 mL | 38-430 mm |
£66.70
Pack of 12 |
In Stock
Log in to see stock. |
||||
10546732
|
Thermo Scientific™
2016-0250 |
250 mL | 38-430 mm |
£118.00
Pack of 12 |
In Stock
Log in to see stock. |
||||
10370811
|
Thermo Scientific™
2016-0500 |
500 mL | 38-430 mm |
£138.00
Pack of 12 |
In Stock
Log in to see stock. |
||||
10617521
|
Thermo Scientific™
2016-1000 |
1 L | 38-430 mm |
£86.05
Pack of 6 |
In Stock
Log in to see stock. |
||||
Description
Easily measure and dispense using Thermo Scientific™ Nalgene™ Square Narrow-Mouth PPCO Bottles with Closure, featuring mouths wider than conventional narrow-mouth bottles. These polypropylene copolymer bottles are graduated, repeatedly autoclavable, translucent, and resistant to alcohols and bases. They are made only from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.
Features:- Square sides allow for good use of space and an easy grip
- Graduated for ease of dispensing
- Linerless caps provide leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
- Autoclavable/Leakproof† when used with Nalgene closures
- Translucent bottles provide visible liquid level
- Useful between 0° and 135°C (32° and 275°F)
- Manufactured from non-toxic materials
Includes:
- Leakproof† polypropylene screw caps
Note:
Remove cap or completely disengage threads before autoclaving. Bottle neck design is protected by US Trademark Reg. No. 2857283. Square bottle with arched shoulders design is protected by US Trademark Reg. No. 2857279.
.
Recycling Information:
Nalgene reusable polypropylene (PP) and polypropylene copolymer (PPCO) bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. These bottles and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene PP and PPCO bottles if services are available in your community.
Specifications
Translucent | |
Leakproof polypropylene screw closures | |
Non-sterile | |
Yes | |
Yes | |
Yes | |
Square | |
Polypropylene | |
No |
Narrow-Mouth Square Bottle | |
PPCO | |
Natural, translucent | |
USP Class VI; FDA - 21 CFR | |
Autoclavable | |
Narrow | |
Square Narrow Mouth Bottle | |
Screw Cap |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.