Learn More
Thermo Scientific™ Nalgene™ Square PET Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
Provide excellent gas barrier properties for storage and shipping of liquid media, buffers and sera with these square bottles.
£175.00 - £287.00
Specifications
Color | Clear |
---|---|
Diameter (Metric) Inner Neck | 28 mm |
Material | PET |
Sterility | Sterile |
Closure Included | Yes |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
15488084
|
Thermo Scientific™
842040-0125 |
125 mL |
£287.00
Pack of 96 |
In Stock
Log in to see stock. |
||||||
10572225
|
Thermo Scientific™
342040-0250 |
250 mL |
£175.00
Pack of 60 |
In Stock
Log in to see stock. |
||||||
15485683
|
Thermo Scientific™
842040-0650 |
650 mL |
£280.00
Pack of 48 |
In Stock
Log in to see stock. |
||||||
15431964
|
Thermo Scientific™
842040-1000 |
1 L |
£259.00
Pack of 24 |
In Stock
Log in to see stock. |
||||||
Description
Provide excellent gas barrier properties for storage and shipping of liquid media, buffers and sera with Thermo Scientific™ Nalgene™ Square PET Media Bottles with Closure. These light-weight bottles are shatter-resistant, sterile and come in a space-saving square design.
Features:- Polyethylene terephthalate with natural high density polyethylene closure
- Molded in graduations and neck ring for use with tamper evident shrink sleeves
- Gamma irradiated to 10-6 Sterility Assurance Level (SAL)
- Non-pyrogenic and non-cytotoxic
- Transparency and reduced permeability to CO2 and O2
- Bottle and closure material are Animal Derived Component (ADC) Free
- Leakproof†
Ordering Information:
Products are packaged in shrink-wrap trays. Trays are double polybagged inside the master carton.
Specifications
Clear | |
PET | |
Yes | |
Narrow | |
Media Bottle |
28 mm | |
Sterile | |
Yes | |
Square | |
38-430 mm |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.