missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Ultra-Low Particulate Media Bottles

Product Code. 16410611
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
2 L
Unit Size:
Pack of 20
Pack of 35
Pack of 48
Pack of 50
Pack of 70
Pack of 72

Product Code. 16410611

Brand: Thermo Scientific™ CEN20190250

Only null left
Add to basket
Edge
Add to basket

Designed for the most critical applications, Nalgene PETG Ultra-Low Particulate Square Media Bottles are platinum certified to contain less than one-third of the USP <788> particulate limits. With exceptionally low particulates and endotoxins, they set the standard for biological and pharmaceutical manufacturing downstream applications.

Products are ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates. They are also well suited for the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.

  • Eliminates costly washing, depyrogenation and autoclaving steps
  • Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and HDPE closures provided
  • Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates
  • Also well suited to the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media.
  • Supported by extensive validation documentation
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
Capacity (English) 8 oz.
Color Clear
Diameter (English) 2.4 in.
Diameter (English) Inner Neck 1.10 in.
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Height (Metric) With Closure 144 mm
Material PETG
No. per Case 48/Cs.
No. per Pack 12/Pk.
Quantity Case of 48
Sterility Sterile
Temperature Range (Metric) -70°C to 70°C
Weight (English) 10.2 lb.
Low Particulate <1/3 USP <788> (Ultralow particulate)
Certifications/Compliance USP Class VI Compliant, Certified Radiation Stabilized (SAL 10-6)
Mouth Narrow
Product Type Media Bottle
Width (English) 12.12 in.
Height (English) 5.75 in.
Closure Size 38-430 mm
Diameter (Metric) 59 mm
Autoclavable Not Autoclavable
Workflow Step Downstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.