Learn More
Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays
Package and ship liquid medias, buffers and sera using these square, heavy-walled, durable bottles that offer excellent gas barrier properties.
£178.00 - £466.00
Specifications
Color | Clear |
---|---|
For Use With (Application) | Storing Media and Sera |
Material | PETG |
Sterility | Non-sterile |
Certifications/Compliance | USP Class VI, FDA Compliance 21 CFR |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | |||||
10561075
|
Thermo Scientific™
322020-0030 |
30 mL | 20-415 mm |
£466.00
Pack of 280 |
In Stock
Log in to see stock. |
|||||
10304293
|
Thermo Scientific™
322020-0060 |
60 mL | 24-415 mm |
£449.00
Pack of 200 |
In Stock
Log in to see stock. |
|||||
10384333
|
Thermo Scientific™
322020-0125 |
125 mL | 38-430 mm |
£352.00
Pack of 96 |
In Stock
Log in to see stock. |
|||||
10140463
|
Thermo Scientific™
322020-0250 |
250 mL | 38-430 mm |
£182.00
Pack of 60 |
In Stock
Log in to see stock. |
|||||
10262953
|
Thermo Scientific™
322020-0500 |
500 mL | 38-430 mm |
£180.00
Pack of 40 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||||
10622634
|
Thermo Scientific™
322020-1000 |
1 L | 38-430 mm |
£271.00
Pack of 24 |
In Stock
Log in to see stock. |
|||||
10222813
|
Thermo Scientific™
322020-2000 |
2 L | 53B |
£178.00
Pack of 12 |
In Stock
Log in to see stock. |
|||||
Description
Package and ship liquid media, buffers and sera using Thermo Scientific™ Nalgene™ PETG Square Media Bottles with Closures. Heavy-walled, durable bottles offer excellent gas barrier properties and are specifically designed for storing media and sera.
- Heavy-walled and durable
- Shatter-resistant
- Provide excellent gas barrier properties
- In shrink-wrap trays
- Molded in graduations
- Clear bottles for easy identification of bottles
- Nonsterile
- Leakproof†
Specifications
Clear | |
PETG | |
USP Class VI, FDA Compliance 21 CFR | |
Yes | |
Square | |
HDPE |
Storing Media and Sera | |
Non-sterile | |
Yes | |
Narrow | |
Media Bottle |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.