missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Septum Closure: Sterile, Shrink-Wrapped Trays Product Code.: 10159742
Add to basket

Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Septum Closure: Sterile, Shrink-Wrapped Trays
GREENER_CHOICE

Product Code. 10159742
60 mL, 24 mm
Click to view available options
Capacity (Metric):
1 L
125 mL
500 Expanded
500 mL
60 mL
Closure Size:
24 mm
38-430 mm
This item is not returnable. View return policy

Product Code. 10159742

Brand: Thermo Scientific™ 3420230060

Add to basket
Add to basket
This item is not returnable. View return policy

Package biological and diagnostic reagents that require aseptic dispensing or addition of supplements by syringe in these durable, break-resistant media bottles.

Thermo Scientific™ Nalgene Square PETG Media Bottles are durable, break-resistant, and feature a septum closure for aseptic syringe dispensing or addition of supplements.

Features:
  • HDPE closure with silicone/PTFE septum provides a PTFE fluid contact surface that is inert and nonstick to ensure fluid integrity.

    • Leakproof†, transparent media bottles
    • Square design saves valuable shelf space
    • Bottles and closures are sterile to 10-6 sterility assurance level

    Ordering Information:

    Packed in shrink-wrapped trays in double-polybagged cartons for cleanroom applications.

    Recommended for:

    Ideal for packaging of biological and diagnostic reagents that require aseptic dispensing or addition of supplements by syringe.

    Note:

    † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 60 mL
Capacity (English) 2 oz.
Color Transparent
Description With septum closure
Diameter (English) 1.6 in.
Diameter (English) Inner Neck 0.71 in.
Diameter (Metric) Inner Neck 18 mm
Material PETG
No. per Case 200
No. per Pack 40
Sterility Sterile
Certifications/Compliance Meet USP Class IV standards. Noncytotoxic.
Closure Included Yes
Graduated Yes
Mouth Wide
Shape Square
Fit for Purpose Sterile
Height (Metric) 82 mm
Product Type Media Bottle
Height (English) 3.23 in.
Closure Size 24 mm
Diameter (Metric) 41 mm
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1422907 Certificate TypeCertificate of Processing Date12/03/2025 Catalog Number342023-0060
Lot Number1381020 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number342023-0060
Lot Number1399782 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number342023-0060
Lot Number1422907 Certificate TypeCertificate of Compliance Date13/02/2025 Catalog Number342023-0060
Lot Number1274505 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number342023-0060
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Septum Closure: Sterile, Shrink-Wrapped Trays >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.