missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Septum Closure: Sterile, Shrink-Wrapped Trays

Catalog Number 10767263
231.76 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.


Click to view available options
Capacity (Metric):
1 L
125 mL
500 Expanded
500 mL
60 mL
Fit for Purpose:
Sterile

Package biological and diagnostic reagents that require aseptic dispensing or addition of supplements by syringe in these durable, break-resistant media bottles.

Thermo Scientific™ Nalgene Square PETG Media Bottles are durable, break-resistant, and feature a septum closure for aseptic syringe dispensing or addition of supplements.

Features:
  • HDPE closure with silicone/PTFE septum provides a PTFE fluid contact surface that is inert and nonstick to ensure fluid integrity.

    • Leakproof†, transparent media bottles
    • Square design saves valuable shelf space
    • Bottles and closures are sterile to 10-6 sterility assurance level

    Ordering Information:

    Packed in shrink-wrapped trays in double-polybagged cartons for cleanroom applications.

    Recommended for:

    Ideal for packaging of biological and diagnostic reagents that require aseptic dispensing or addition of supplements by syringe.

    Note:

    † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

TRUSTED_SUSTAINABILITY
Capacity (Metric) 1 L
Capacity (English) 33.8 oz.
Color Transparent
Description With septum closure
Diameter (English) 3.7 in.
Diameter (English) Inner Neck 1.1 in.
Diameter (Metric) Inner Neck 28 mm
Material PETG
No. per Case 24
No. per Pack 12
Sterility Sterile
Certifications/Compliance Meet USP Class IV standards. Noncytotoxic.
Closure Included Yes
Graduated Yes
Mouth Wide
Shape Square
Fit for Purpose Sterile
Height (Metric) 220 mm
Product Type Square Bottle
Height (English) 8.66 in.
Closure Size 38-430 mm
Diameter (Metric) 94 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'