missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
GREENER_CHOICE

Product Code. 10542111
Click to view available options
Capacity (Metric):
60 mL
175 mL
250 mL
500 mL
1 L
Closure Size:
28 mm
38 mm
43 mm
53 mm
63 mm
missing translation for 'unitSize'
Pack of 12
Pack of 6

Code produit. 10542111

missing translation for 'mfr': Thermo Scientific™ 21100032

Il en reste null
Ajouter au panier
Edge
Ajouter au panier

Simplify cleaning, sample filling and retrieval with these space-efficient, wide-mouth PPCO bottles, which are contact-clear, autoclavable and resistant to most acids, alcohols and bases.

Save valuable lab space with the square design of Thermo Scientific™ Nalgene™ Wide-Mouth Square Polypropylene Copolymer Bottles. Wide-mouth design allows easy cleaning, sample filling and retrieval. Made from only high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.

Features:
  • Resistant to acids, alcohols and bases
  • Contact-clear bottle provides visible liquid level
  • Square design provides a large area for labeling
  • Linerless polypropylene closures provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Autoclavable

Note:

Completely disengage threads or remove cap before autoclaving.

TRUSTED_SUSTAINABILITY

Spécification

Capacity 32 oz., 1000 mL
Capacity (Metric) 1 L
Capacity (English) 32 oz.
Color Translucent
Description Wide-Mouth Square Bottle
Diameter (English) 3.7 in.
Diameter (English) Inner Neck 2 in.
Diameter (Metric) Inner Neck 53 mm
Material PPCO
No. per Case 24
No. per Pack 6
Sterility Non-sterile
Closure Color Natural, translucent
Certifications/Compliance USP Class VI; FDA - 21 CFR
Closure Included Yes
Graduated No
Mouth Wide
Shape Square
Height (Metric) 181 mm
Product Type Square Wide Mouth Bottle
Width (English) 3.72 in.
Height (English) 7.1 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 63 mm
Diameter (Metric) 95 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Autoclavable Autoclavable
Afficher plus Afficher moins

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis