Learn More
Thermo Scientific™ Nalgene™ Stainless-Steel Forceps, bent tip
Nalgene Stainless-Steel Forceps are available in bent or straight-tip styles. Bent-tip style is designed for easy removal and manipulation of filter membranes.
Brand: Thermo Scientific™ DS0399-0001
31.80 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : A0
Additional Details : Weight : 0.04320kg
Description
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
- Stainless steel
- Self-closing (bent tip)
- Serrated gripping area
- Autoclavable
Specifications
Bent, Round, Serrated | |
Stainless Steel | |
Stainless-Steel Forceps, bent tip | |
Self-closing Bent |
1 | |
Forceps | |
Ea. |