missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene Stopcock Assembly for Acrylic Burets
GREENER_CHOICE

For use with 25 mL Acrylic Nalgene Break-Resistant Burets.

Brand:  Thermo Scientific™ 713650-1025

Product Code. 10642102

  • £230.00 / Pack of 2
Estimated Shipment: 11-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For use with: Nalgene 25mL Acrylic Buret (Cat. No. 36500025)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Self Lubricating
PTFE
25 mm
Stopcock Assy Only 3650-0025 25mL
0.98 in.
Nalgene 25mL Acrylic Buret (36500025)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.