missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ Centrifuge Bottles made with Teflon™ fluoropolymer, 250mL Product Code.: 10549681
Add to basket

Thermo Scientific™ Nalgene™ Centrifuge Bottles made with Teflon™ fluoropolymer, 250mL
GREENER_CHOICE

Product Code. 10549681
250 mL, 4000 x g
Click to view available options
Nominal Capacity:
250 mL
Max. Force:
4000 x g
This item is not returnable. View return policy

Product Code. 10549681

Brand: Thermo Scientific™ 31270250

Add to basket
Add to basket
This item is not returnable. View return policy

Perform low-speed, lipid and phenol extractions, digestions and precipitations with Thermo Scientific™ Nalgene™ Centrifuge Bottles made with Teflon™ fluoropolymer.

Perform low-speed, lipid and phenol extractions, digestions and precipitations with Thermo Scientific™ Nalgene™ Centrifuge Bottles made with Teflon™ fluoropolymer. The perfect fit with Thermo Scientific™ Fiberlite™ carbon fiber rotors, they are molded of translucent FEP that resists most acids, bases and organic solvents.

Features:
  • FEP provides excellent chemical resistance
  • Use in temperatures from -100° to +150°C
  • For proper performance, must be filled to 100% of total capacity
  • For added leakproof† asssurance at maximum speeds or when spinning hazardous materials use with PP sealing caps assemblies (DS3131-0038)

Requires:

When spinning 250mL bottle in a Sorvall GSA rotor, use of the Nalgene centrifuge bottle adaptor (DS3125-0250) is required.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Color Clear
Description Teflon FEP Bottle with Tefzel ETFE closure
Material FEP
No. per Case 6
No. per Pack 1
Nominal Capacity 250 mL
Graduated No
Max. Force 4000 x g
Shape Round
Height (Metric) 126.5 mm
Product Type Centrifuge Bottle
Closure Material Tefzel ETFE
Diameter (Metric) 60.0 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Show More Show Less

Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.

Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Centrifuge Bottles made with Teflon™ fluoropolymer, 250mL >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.