Learn More
Thermo Scientific™ Nalgene™ Drop-Dispensing Bottle made with Teflon™ fluoropolymer, Dropping Closure and Cap made with Tefzel™
Description
Dispense strong acids/alkalis, reagents and solvents with Thermo Scientific™ Nalgene™ Drop-Dispensing Bottle made with Teflon™ fluoropolymer, Dropping Closure and Cap made with Tefzel™, featuring excellent chemical resistance for dispensing virtually any chemical. Tethered spout cap snaps securely into place and won't pop off or leak when the closed bottle is squeezed. Usable over a wide temperature range -105°C to +150°C. Leakproof† when used with provided Nalgene closure and autoclavable.
- Dispenses only one drop at a time for dependable delivery
- Convenient one-handed operation frees up your other hand
- Transparent so you can easily view liquid levels
- Includes ETFE dropping closure and cap
Ordering Information:
One per package; case of four.
- Environmental benefits include:
- Renewable Energy
Specifications
Specifications
Capacity (Metric) | 30 mL |
Capacity (English) | 1 oz. |
Color | White |
Description | Teflon FEP Drop-Dispensing Bottle with Dropping Closure and Cap |
Material | FEP |
No. per Case | 4 |
No. per Pack | 1 |
Certifications/Compliance | USP Class VI |
Closure Included | Yes |
Autoclavable | Autoclavable |
Show More |
Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.
Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.