Learn More
Thermo Scientific™ Nalgene™ Torque Wrench Fittings for Biotainer™ Closures
Assure accurate torque readings when applying closures with these torque wrench fittings, designed specifically for BiotainerTM closures.
£158.00 - £175.00
Specifications
Product Type Fittings
Drive Size 1/4 in.
Material Epoxy
No. per Pack 1 Ea.
View More Specs
Products 2
Product Code Brand Closure Size Price Quantity & Availability  
Product Code Brand Closure Size Price Quantity & Availability  
11740094
View Documents
Thermo Scientific™
2595-3038
38 to 430 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10262700
View Documents
Thermo Scientific™
2595-9048
48 to 430 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Assure accurate torque readings when applying closures with Thermo Scientific™ Nalgene™ Torque Wrench Fittings for Biotainer™ Closures. Proper torquing of Nalgene closures is critical to leakproof† sealing.
  • Epoxy
  • Specifically designed for Nalgene Biotainer closures
Compatible with:
Thermo Scientific Nalgene Torque Wrenches (2195-6169 and -6177)
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Specifications
Fittings
1/4 in.
Epoxy
1 Ea.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Torque Wrench Fittings for Biotainer™ Closures