missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Translucent Amber PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules
GREENER_CHOICE

Protect against leakage, breakage, and contamination with Nalgene Sterile PETG Serum Vials. For use with lined continuous thread closures.

Brand:  Thermo Scientific™ 342033-0010

524.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10039492

  • £664.41 / Pack of 1260
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Protect against leakage, breakage, and contamination with Thermo Scientific™ Nalgene Sterile PETG Serum Vials. Made for use with lined continuous thread closures.

  • PETG
  • Non-cytotoxic
  • Non-pyrogenic
  • Translucent amber
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Sterile to 10-6 SAL
  • Leakproof† with Nalgene lined continuous thread closures (Cat. No. 342158)

Ordering Information:

Serum Vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 342158).

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.

Note: These tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Serum Vials
Amber Translucent
PETG
10 mL
252
Nalgene
Lined Continuous-thread Closure (Sold Separately)
1260 Cases, 252 Pk.
Serum Insert Vial
Lined Continuous-Thread Closure
11 mm
24 mm
Sterile
1260
1/3 oz.
10 mL
54 mm
ISO 13485:2016
20-415
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1271340 Certificate TypeCertificate of Compliance Date11/12/2024 Catalog Number342033-0010
Lot Number1301093 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number342033-0010
Lot Number1305148 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number342033-0010
Lot Number1386727 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number342033-0010
Lot Number1382809 Certificate TypeCertificate of Processing Date04/12/2024 Catalog Number342033-0010
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Translucent Amber PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.