missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Tubing Y-Type Connectors

Maximize liquid flow with non-tapered hose barbed Y-Type Connectors, molded of polypropylene resin, free of animal derived components.

Brand:  Thermo Scientific™ 6152-0250

50.82 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.



Code : 55

Additional Details : Weight : 0.01980kg

 View more versions of this product

Product Code. 10186070

  • £59.25 / Pack of 12
In Stock
to see stock.
Add to basket

Description

Description

Maximize liquid flow with non-tapered hose barbed Thermo Scientific™ Nalgene™ Tubing Y-Type Connectors, molded of polypropylene resin, free of animal derived components. Products are double-bagged to maintain cleanliness.

  • Polypropylene construction
  • Autoclavable
Specifications

Specifications

Tubing Connector
Autoclavable
1/4 in.
1/4 in. I.D.
Y-Type Connector
Polypropylene
6.35 mm
12/Pk., 72/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.