Learn More
Thermo Scientific™ Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
GREENER_CHOICE
Securely hold a full load of test tubes or centrifuge tubes for 13, 16, 20, 25 and 30mm diameter sizes with these strong, durable racks.
£23.30 - £62.55
Specifications
Product Type Test Tube Rack
Autoclavable Autoclavable
Material RESMER
Shape Rectangular
View More Specs
Products 30
Product Code Brand Array Color For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Array Color For Use With (Equipment) Price Quantity & Availability  
10603161
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0513
6 x 12 Red 13mm Tubes
£52.35
Each
In Stock
to see stock.
Edge
Add to basket
10149970
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0313
6 x 12 Blue 13mm Tubes
£53.20
Each
In Stock
to see stock.
Edge
Add to basket
10180351
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0413
6 x 12 Green 13mm Tubes
£52.55
Each
In Stock
to see stock.
Add to basket
10333461
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0213
6 x 12 Yellow 13mm Tubes
£53.00
Each
In Stock
to see stock.
Add to basket
10634481
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0113
6 x 12 Orange 13mm Tubes
£53.20
Each
In Stock
to see stock.
Add to basket
10693151
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0013
6 x 12 White 13mm Tubes
£54.10
Each
In Stock
to see stock.
Add to basket
10435071
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0516
6 x 12 Red 16mm Tubes
£53.85
Each
In Stock
to see stock.
Edge
Add to basket
10098210
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0316
6 x 12 Blue 16mm Tubes
£55.20
Each
In Stock
to see stock.
Edge
Add to basket
10495061
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0416
6 x 12 Green 16mm Tubes
£51.45
Each
In Stock
to see stock.
Edge
Add to basket
10343461
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0216
6 x 12 Yellow 16mm Tubes
£51.75
Each
In Stock
to see stock.
Add to basket
10717961
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0116
6 x 12 Orange 16mm Tubes
£53.50
Each
In Stock
to see stock.
Edge
Add to basket
10281761
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0016
6 x 12 White 16mm Tubes
£55.55
Each
In Stock
to see stock.
Edge
Add to basket
10519081
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0520
4 x 10 Red 20mm Tubes
£54.35
Each
In Stock
to see stock.
Edge
Add to basket
10323701
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0320
4 x 10 Blue 20mm Tubes
£55.15
Each
In Stock
to see stock.
Edge
Add to basket
10769491
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0420
4 x 10 Green 20mm Tubes
£53.85
Each
In Stock
to see stock.
Edge
Add to basket
10604101
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0220
4 x 10 Yellow 20mm Tubes
£52.80
Each
In Stock
to see stock.
Add to basket
10169970
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0120
4 x 10 Orange 20mm Tubes
£54.05
Each
In Stock
to see stock.
Add to basket
10291761
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0020
4 x 10 White 20mm Tubes
£31.05
Each
In Stock
to see stock.
Edge
Add to basket
10663721
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0525
4 x 10 Red 25mm Tubes
£23.30
Each
In Stock
to see stock.
Add to basket
10415411
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0325
4 x 10 Blue 25mm Tubes
£59.05
Each
In Stock
to see stock.
Edge
Add to basket
10231671
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0425
4 x 10 Green 25mm Tubes
£24.50
Each
Estimated Shipment: 08-09-2025
to see stock.
Add to basket
10602401
GREENER_CHOICE
View Documents
Thermo Scientific™
5970-0225
4 x 10 Yellow 25mm Tubes
£24.06
Each
In Stock
to see stock.
Add to basket
10738151
GREENER_CHOICE
View Documents
Thermo Scientific™
5970-0125
4 x 10 Orange 25mm Tubes
£23.30
Each
Estimated Shipment: 16-09-2025
to see stock.
Add to basket
10474101
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0025
4 x 10 White 25mm Tubes
£34.95
Each
In Stock
to see stock.
Edge
Add to basket
10241861
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0530
3 x 8 Red 30mm Tubes
£58.55
Each
In Stock
to see stock.
Edge
Add to basket
10778721
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0330
3 x 8 Blue 30mm Tubes
£62.55
Each
In Stock
to see stock.
Edge
Add to basket
10602971
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0430
3 x 8 Green 30mm Tubes
£60.60
Each
In Stock
to see stock.
Edge
Add to basket
10038180
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0230
3 x 8 Yellow 30mm Tubes
£58.15
Each
In Stock
to see stock.
Edge
Add to basket
10242101
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0130
3 x 8 Orange 30mm Tubes
£60.60
Each
In Stock
to see stock.
Edge
Add to basket
10644101
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
5970-0030
3 x 8 White 30mm Tubes
£58.70
Each
In Stock
to see stock.
Edge
Add to basket
Description
Description
Securely hold a full load of test tubes or centrifuge tubes for 13, 16, 20, 25 and 30mm diameter sizes with Thermo Scientific™ Nalgene™ Unwire™ Test Tube Racks. Molded in a single piece for exceptional strength and durability, these racks have no coating to chip, peel or rust.
  • Smooth finish leaves fewer places for contaminants to lodge; no welds to break, no sharp edges.
  • Endplates accept large labels for easy identification of samples.
  • Higher specific gravity than water so it stays submerged in a water bath and won't float.
  • Withstands most lab chemicals and temperatures up to 121°C for a variety of lab procedures.
  • Available in six bright colors to help keep samples organized.
  • 15mL conical tubes fit 20mm racks; 50mL conical fit 30mm racks.
  • No assembly is required; rack is ready to use.
Please recycle the corrugated cartons and LDPE bags in which your Nalgene products are packaged.
Specifications
Specifications
Test Tube Rack
Autoclavable
RESMER
Rectangular
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology