missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Utility Funnels
GREENER_CHOICE

Replace glass lab funnels with these utility funnels, which are an excellent shatter-proof alternative.

Brand:  Thermo Scientific™ 4256-0638

244.80 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10440723

  • £334.00 / Pack of 12
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

Replace glass lab funnels with Thermo Scientific™ Nalgene™ Utility Funnels. These funnels are chemically resistant to a wide variety of lab chemicals and reagents and are shatterproof making them a valuble asset to lab safety programs.

  • 4256-0234 and 4256-0414 are made from shatter-proof, autoclavable polypropylene.
  • 4256-0638 has a large throat opening for faster transfer of materials and is made from shatter-proof high-density polyethylene (Not autoclavable).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

150 mm
48 mm
175 mm
Funnel Utility HDPE 162mm
12/Pk.; 72/Cs.
1059 mL
HDPE
Not Autoclavable
66 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.