missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Utility Funnels
Replace glass lab funnels with these utility funnels, which are an excellent shatter-proof alternative.
Brand: Thermo Scientific™ 4256-0414
38.01 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Replace glass lab funnels with Thermo Scientific™ Nalgene™ Utility Funnels. These funnels are chemically resistant to a wide variety of lab chemicals and reagents and are shatterproof making them a valuble asset to lab safety programs.
- 4256-0234 and 4256-0414 are made from shatter-proof, autoclavable polypropylene.
- 4256-0638 has a large throat opening for faster transfer of materials and is made from shatter-proof high-density polyethylene (Not autoclavable).
Specifications
98 mm | |
18 mm | |
117 mm | |
Funnel Utility PP 108mm | |
12/Pk.; 144/Cs. |
270 mL | |
Polypropylene | |
Autoclavable | |
37 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction