missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Acetone
Distilled Water
Isopropanol
Sodium Hypochlorite
Description
Keep hazardous volatile liquids off the lab bench with Thermo Scientific™ Nalgene™ Vented Unitary Right-To-Know LDPE Wash Bottles, featuring a unique vent system. Unitary bottle prevents contamination when filling the bottle; no draw tube assembly to pick up contamination off the lab bench. A strong, steady, no sputter stream of liquid is dispensed when the wash bottle is squeezed. Marked with four color, NFPA diamonds, GHS compliant hazard symbols, and right-to-know chemical information. Choose from seven chemical markings.
- Bottle and tubulation molded in one piece to prevent chemical splash when removing closure and contamination when filling
- Unique 38mm colored vented closure designed to prevent dripping
- Sturdy polypropylene closures (HDPE on Sodium Hypochloride bottles)
- Color-coded bar corresponds to closure, preventing cross-contamination
- Prelabeled with ICS name and formula, as well as NFPA symbols for chemical handling, hazard, care and storage of six commonly-used chemicals
- Leakproof†
Specifications
Specifications
Capacity | 16 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Translucent |
Description | RTK Wash Bottle |
Material | LDPE |
No. per Case | 24 |
No. per Pack | 4 |
Closure Color | Natural |
Unitary | Yes |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'