missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Wide-Mouth HDPE IP2 Bottles with Closure: Bulk Pack
GREENER_CHOICE

Product Code. 10484933
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
Closure Size:
38-415 mm
43-415 mm
53-415 mm
63-415 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Store and ship liquid reagents with these leakproof, wide-mouth bottles with closure that meet ICAO and IATA requirements.

Store and ship liquid reagents with Thermo Scientific™ Nalgene™ Wide-Mouth HDPE IP2 Bottles with Closure. Leakproof† when used with Nalgene closures and constructed of durable high-density polyethylene, these safe, economical bottles are recommended for those designing, assembling and certifying their own combination packaging.

  • HDPE bottles with PP Closures, bulk
  • Wide-mouth
  • Complies with IATA and ICAO regulations
  • For combination packaging
  • Leakproof

Ordering Information:

Bottles and closures are bulk packed in separate polybags within the same carton

Recommended for:

Storage and shipment of liquid reagents. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 16 oz., 500 mL
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color White
Description IP2 Wide-Mouth Bottles
Diameter (Metric) Inner Neck 44 mm
Diameter (Metric) Inner 44 mm
For Use With (Application) Storage and Shipment of Liquid Reagents
Height (Metric) With Closure 168 mm
Material HDPE
No. per Case 125
No. per Pack 125
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27 (c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9.
Closure Included Yes
Mouth Wide
Shape Round
Height (Metric) 168 mm
Product Type IP2 Bottle
Height (English) 6.6 in.
Closure Material Polypropylene
Closure Size 53-415 mm
Diameter (Metric) 73 mm
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'