missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Wide-Mouth Lab Quality HDPE Bottles
GREENER_CHOICE

Product Code. 10569831
125 mL, 38 mm
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
1500 mL
Closure Size:
28 mm
38 mm
43 mm
53 mm
63 mm

Perform countless field applications utilizing these general-purpose, wide-mouth, lab quality HDPE bottles.

Utilize Thermo Scientific™ Nalgene™ Wide-Mouth Lab Quality HDPE Bottles for countless field applications. These containers exhibit excellent chemical resistance to most acids, bases and alcohols and are suitable for freezer use to -100°C and are Leakproof† when used with Nalgene closures. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

Features:

  • Excellent chemical resistance to most corrosives make these bottles versatile and indispensable in your lab
  • Ideal for storing, shipping, and packaging liquids — uniform walls provide durability and resistance to splitting or punctures
  • Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
  • Participates in My Green Labs' ACT Label program for Accountability, Consistency and Transparency around product sustainability

Includes:

Polypropylene screw closure

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Capacity 4 oz., 125 mL
Capacity (Metric) 125 mL
Capacity (English) 4 oz.
Color Translucent
Description Lab Quality Wide-Mouth Bottle
Diameter (Metric) Inner Neck 28 mm
For Use With (Application) Storage and Shipping of Small Volume, Light Sensitive, Valuable Reagents
Material HDPE
No. per Case 72
No. per Pack 12
Sterility Non-sterile
Closure Color Natural
Neck Finish 38 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Disposable Reusable
Mouth Wide
Shape Round
ACT Label Yes
Height (Metric) 99 mm
Product Type Wide Mouth Lab Quality Bottle
Height (English) 11.1 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 38 mm
Diameter (Metric) 50 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Wide-Mouth Lab Quality HDPE Bottles >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.