missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naloxone hydrochloride, 98%, Thermo Scientific™
Click to view available options
Quantity:
100mg
500mg
1g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Naloxone hydrochloride the hydrochloride salt of the anti-opioid naloxone
• Naloxone reverses characteristic opioid effects including euphoria, bradycardia, sedation, and respiratory depression
Application
• Naloxone is a fast-acting inverse agonist of the mu-opioid receptor that can be used to competitively inhibit the binding of opioid compounds
• Naloxone is appropriate for use in in vivo and in vitro studies of opioid receptor signaling
Specifications
Specifications
Quantity | 100mg |
Merck Index | 14,6362 |
Solubility Information | Soluble in water (73mg/ml),ethanol (3.3mg/ml),methanol (50mg/ml) and dimethyl sulfoxide (73mg/ml). Insoluble in ether |
Formula Weight | 363.84 |
Percent Purity | 98% |
Chemical Name or Material | Naloxone hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'